2-Ethoxyphenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Ethoxyphenylisothiocyanate

TL00819.1 TL00819EA TL00819.50
MAYBTL00819.1EA 0 EUR
MAYBTL00819.1 MAYBTL00819EA MAYBTL00819.50
2-Ethoxyphenyl isothiocyanate
2-Ethoxyphenyl isothiocyanate
Formule: C₉H₉NOS
MW: 179.24 g/mol
MDL Nummer: MFCD00046793
CAS nummer: 23163-84-0
EINECS: 245-468-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR