3,4-Dimethoxyphenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

S03458.1 S03458EA S03458.50
MAYBS03458.1EA 0 EUR
MAYBS03458.1 MAYBS03458EA MAYBS03458.50
3,4-Dimethoxyphenyl isothiocyanate
3,4-Dimethoxyphenyl isothiocyanate
Formule: C₉H₉NO₂S
Kookpunt: 171…172 °C (7 mmHg)
MDL Nummer: MFCD00041075
CAS nummer: 33904-04-0
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR