3,5-Dimethoxyphenyl isothiocyanate
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1,3-Dimethoxy-5-isothiocyanatobenzene
TL00503.1
TL00503.10
TL00503.50
MAYBTL00503.1EA
0
EUR
MAYBTL00503.1
MAYBTL00503.10
MAYBTL00503.50
3,5-Dimethoxyphenyl isothiocyanate
3,5-Dimethoxyphenyl isothiocyanate
Formule:
C₉H₉NO₂S MW: 195.24 g/mol Kookpunt: 130…132 °C (3 mmHg) Smeltpunt: 51 |
MDL Nummer:
MFCD00041077 CAS nummer: 104968-58-3 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...