m-Anisidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine

SB01292.1 SB01292.10 SB01292.50
MAYBSB01292.1EA 0 EUR
MAYBSB01292.1 MAYBSB01292.10 MAYBSB01292.50
m-Anisidine
m-Anisidine
Formule: C₇H₉NO
MW: 123.15 g/mol
Kookpunt: 251
Smeltpunt: –2 °C
Dichtheid: 1.1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007783
CAS nummer: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1 III
Merck Index: 13,00670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR