Sulfanilzuur 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
20674.231EA
75.8
EUR
20674.231
Sulfanilzuur 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Sulfanilzuur
Formule:
4-(H₂N)C₆H₄SO₃H MW: 173,19 g/mol Kookpunt: 363 °C (1013 hPa) Smeltpunt: 279 °C Dichtheid: 1,485 g/cm³ (25 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007886 CAS nummer: 121-57-3 EINECS: 204-482-5 Merck Index: 13,09012 |
Specification Test Results
Assay | 98.0 - 102.0 % |
Ignition residue (SO4) | Max. 100 ppm |
Insolubility in Na2CO3 solution | Max. 0.02 % |
Water | Max. 0.7 % |
Cl (Chloride) | Max. 20 ppm |
NO2 (Nitrite) | Max. 0.5 ppm |
SO4 (Sulphate) | Max. 100 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...