4-Methoxythiophenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00258.1 TL00258.10 TL00258.50
MAYBTL00258.1EA 0 EUR
MAYBTL00258.1 MAYBTL00258.10 MAYBTL00258.50
4-Methoxythiophenol
4-Methoxythiophenol
Formule: C₇H₈OS
Kookpunt: 228 °C (1013 hPa)
Smeltpunt: 42 °C
Dichtheid: 1,1397 g/cm³ (20 °C)
MDL Nummer: MFCD00004849
CAS nummer: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR