4-Hydroxythiophenol 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4-Hydroxybenzenethiol, p-Hydroxythiophenol, 1-Hydroxy-4-mercaptobenzene, 4-Mercaptophenol, 4-Hydroxyphenyl mercaptan, 4-Hydroxybenzene-1-thiol

370630050 370630250
ACRO370630050EA 70.1 EUR
ACRO370630050 ACRO370630250
4-Hydroxythiophenol 99%
4-Hydroxythiophenol
Formule: C₆H₆OS
Kookpunt: 149…150 °C (25 mmHg)
Smeltpunt: 33…35 °C
Vlampunt: 110 °C
MDL Nummer: MFCD00004850
CAS nummer: 637-89-8
EINECS: 211-307-6
UN: 2923

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR