Benzenesulphonyl chloride ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Benzene sulphochloride
A10849.0B
A10849.0I
A10849.30
A10849.0BEA
66.1
EUR
A10849.0B
A10849.0I
A10849.30
Benzenesulphonyl chloride ≥98%
Benzenesulphonyl chloride
Purity: 98%
CAS No.: 98-09-9
Molecular Formula: C6H5ClO2S
Formula Weight: 176.62
Flash Point: 132°(270°F)
Refractive Index: 1.5525
Odour: Irritating
Melting Point: 14-15°
Boiling Point: 252°
Beilstein Reference: 606926
Merck Reference: 14,1072
UN No.: 2225
MDL No.: MFCD00007426
Formule:
C₆H₅ClO₂S MW: 176.62 g/mol Kookpunt: 252 °C Smeltpunt: 14…15 °C Dichtheid: 1.377 Vlampunt: 132 °C (270 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007426 CAS nummer: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8 III Merck Index: 13,01072 |