Benzenesulphonohydrazide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Benzenesulphonic acid hydrazide, Benzenesulphonyl hydrazide

B23971.14 B23971.22 B23971.36
B23971.14EA 44 EUR
B23971.14 B23971.22 B23971.36
Benzenesulphonohydrazide ≥98%
Benzenesulphonohydrazide

Purity: 98% CAS No.: 80-17-1 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 101-105° Beilstein Reference: 640079 Merck Reference: 14,7598 UN No.: 3226 MDL No.: MFCD00007583

Formule: C₆H₈N₂O₂S
MW: 172.21 g/mol
Smeltpunt: 101…105 °C
Dichtheid: 1.369 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007583
CAS nummer: 80-17-1
EINECS: 201-255-2
UN: 3226
ADR: 4.1 II
Merck Index: 12,07757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR