Benzenesulphonic acid ≥94%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A16148.36 A16148.0E
A16148.36EA 47 EUR
A16148.36 A16148.0E
Benzenesulphonic acid ≥94%
Benzenesulphonic acid
Purity: 94% CAS No.: 98-11-3 Molecular Formula: C6H6O3S Formula Weight: 158.18 Flash Point: 110°(230°F) Melting Point: ca 45-50° Beilstein Reference: 742513 Merck Reference: 14,1070 UN No.: 2585 MDL No.: MFCD00011689
Formule: C₆H₆O₃S
MW: 158.18 g/mol
Kookpunt: 319
Smeltpunt: ∼ 45…50 °C
Dichtheid: 1.32
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011689
CAS nummer: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8 III
Merck Index: 13,01070