4-Formylfenylboronzuur

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

SB02099.1 SB02099.10 SB02099.50
MAYBSB02099.1EA 0 EUR
MAYBSB02099.1 MAYBSB02099.10 MAYBSB02099.50
4-Formylfenylboronzuur
4-Formylfenylboronzuur
Formule: C₇H₇BO₃
MDL Nummer: MFCD00151823
CAS nummer: 87199-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR