ω-Bromo-3,4-dichloroacetophenone ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one, 2-Bromo-1-(3,4-dichlorophenyl)ethanone, 2-Bromo-3',4'-dichloroacetophenone, 3,4-Dichlorophenacyl bromide, α-Bromo-3,4-dichloroacetophenone
A13365.06
A13365.14
A13365.22
A13365.06EA
42.3
EUR
A13365.06
A13365.14
A13365.22
ω-Bromo-3,4-dichloroacetophenone ≥98%
ω-Bromo-3,4-dichloroacetophenone
Purity: 98%
CAS No.: 2632-10-2
Molecular Formula: C8H5BrCl2O
Formula Weight: 267.94
Storage Temperature: 2°C to 8°C
Melting Point: 55-58°
Beilstein Reference: 511981
UN No.: 1759
MDL No.: MFCD00051581
Formule:
C₈H₅BrCl₂O Smeltpunt: 55…58 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00051581 CAS nummer: 2632-10-2 ADR: 8,III |