2-Tolualdehyde
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
o-Tolualdehyde, 2-Methylbenzaldehyde
SB01011.1
SB01011.10
SB01011.50
MAYBSB01011.1EA
0
EUR
MAYBSB01011.1
MAYBSB01011.10
MAYBSB01011.50
2-Tolualdehyde
2-Tolualdehyde
Formule:
C₈H₈O Kookpunt: 201 °C (1013 hPa) Smeltpunt: –35 °C Dichtheid: 1,0324 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003338 CAS nummer: 529-20-4 EINECS: 208-452-2 Merck Index: 12,09665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...