3-(Chloromethyl)benzoyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
314430050 314430250
ACRO314430050EA 54.6 EUR
ACRO314430050 ACRO314430250
3-(Chloromethyl)benzoyl chloride 98%
3-(Chloromethyl)benzoyl chloride
Formule: C₈H₆Cl₂O
Kookpunt: 149…150 °C (20 mmHg)
Smeltpunt: 4 °C
Dichtheid: 1,33 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000682
CAS nummer: 63024-77-1
EINECS: 263-799-7
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %