3-Chloro-4-metihybenzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00701.1 TL00701.10
MAYBTL00701.1EA 0 EUR
MAYBTL00701.1 MAYBTL00701.10
3-Chloro-4-metihybenzamide
3-Chloro-4-metihybenzamide
Formule: C₈H₈ClNO
CAS nummer: 24377-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR