Safety shoes, lace-up, suXXeed® Offroad, 62743, Heckel
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6274347 6274348 6274339 6274340 6274341 6274342 6274343 6274344 6274336 6274337 6274338 6274345 6274346
UVEX6274347EA 0 EUR
UVEX6274347 UVEX6274348 UVEX6274339 UVEX6274340 UVEX6274341 UVEX6274342 UVEX6274343 UVEX6274344 UVEX6274336 UVEX6274337 UVEX6274338 UVEX6274345 UVEX6274346
Safety shoes, lace-up, suXXeed® Offroad, 62743, Heckel
Schoenen en laarzen
Metal-free, flexible, lightweight safety shoe made from breathable suede leather upper plus a double-desity PU2D sole, highly slip resistant and resistant to oils and hydrocarbons.
  • Quick drying Climate Airmesh lining, very resistant to abrasion, also very good water vapour absorption
  • Removable hygienic insole with abrasion resistant textile top layer
  • Metal-free protective toecap (200 J)
  • Metal-free, puncture resistant midsole
Suitable for warehousing and logistics, services, industry, hot and dry environments.
Colour: Brown/black
Certificaten: EN ISO 20345: 2011; S3, C1, SRC