2,5-Dichlorobenzylamine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (2,5-Dichlorophenyl)methanamine

TL00182DA TL00182EA
MAYBTL00182DAEA 0 EUR
MAYBTL00182DA MAYBTL00182EA
2,5-Dichlorobenzylamine
2,5-Dichlorobenzylamine
Formule: C₇H₇Cl₂N
MW: 176.04 g/mol
Kookpunt: 124…126 °C (13 mmHg)
Dichtheid: 1.2334 g/cm³ (20 °C)
MDL Nummer: MFCD00052391
CAS nummer: 10541-69-2
UN: 2922
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR