2-Chlorobenzyl alcohol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01129.1 SB01129.10 SB01129.50
MAYBSB01129.1EA 0 EUR
MAYBSB01129.1 MAYBSB01129.10 MAYBSB01129.50
2-Chlorobenzyl alcohol
2-Chlorobenzyl alcohol
Formule: C₇H₇ClO
Kookpunt: 227 °C (1013 hPa)
Smeltpunt: 68…71 °C
MDL Nummer: MFCD00004604
CAS nummer: 17849-38-6
EINECS: 241-801-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR