Benzyl cyanide ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Phenylacetonitrile
A10694.0E
A10694.22
A10694.36
A10694.0EEA
226
EUR
A10694.0E
A10694.22
A10694.36
Benzyl cyanide ≥98%
Benzyl cyanide
Purity: 98+%
CAS No.: 140-29-4
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 101°(213°F)
Physical Form: Liquid
Density: 1.015
Refractive Index: 1.5230
Appearance: Colourless
Melting Point: -24°
Boiling Point: 234°
Beilstein Reference: 385941
Merck Reference: 14,1131
UN No.: 2470
MDL No.: MFCD00001894
Formule:
(C₆H₅)CH₂CN Kookpunt: 234 °C Smeltpunt: –24 °C Dichtheid: 1,015 Vlampunt: 101 °C (213 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001894 CAS nummer: 140-29-4 EINECS: 205-410-5 UN: 2470 ADR: 6.1,III Merck Index: 14,1131 |