4-Ethylbenzoyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4-Ethylbenzene-1-carbonyl chloride, 4-Ethylbenzoylchloride

BTB10319.1 BTB10319EA BTB10319.50
MAYBBTB10319.1EA 0 EUR
MAYBBTB10319.1 MAYBBTB10319EA MAYBBTB10319.50
4-Ethylbenzoyl chloride
4-Ethylbenzoyl chloride
Formule: C₉H₉ClO
Kookpunt: 235…236 °C (1013 hPa)
Smeltpunt: 42…43 °C
Dichtheid: 1,14 g/cm³ (20 °C)
MDL Nummer: MFCD00000697
CAS nummer: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR