4-Heptylbenzoyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SPB07640.1 SPB07640.10 SPB07640.50
MAYBSPB07640.1EA 0 EUR
MAYBSPB07640.1 MAYBSPB07640.10 MAYBSPB07640.50
4-Heptylbenzoyl chloride
4-Heptylbenzoyl chloride
Formule: C₁₄H₁₉ClO
Kookpunt: 111 °C (1013 hPa)
Dichtheid: 1,002 g/cm³ (20 °C)
MDL Nummer: MFCD00000702
CAS nummer: 50606-96-7
EINECS: 256-648-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR