4-Toluoyl chloride ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A15581.0E
A15581.22
A15581.36
A15581.0EEA
364
EUR
A15581.0E
A15581.22
A15581.36
4-Toluoyl chloride ≥99%
4-Toluoyl chloride
Purity: 99%
CAS No.: 874-60-2
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 82°(179°F)
Physical Form: Liquid
Density: 1.169
Refractive Index: 1.5530
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Odour: Pungent
Melting Point: -2°
Boiling Point: 225-227°
Beilstein Reference: 471492
UN No.: 3265
MDL No.: MFCD00000696
Formule:
C₈H₇ClO Kookpunt: 225…227 °C Smeltpunt: –2 °C Dichtheid: 1,169 Vlampunt: 82 °C (179 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000696 CAS nummer: 874-60-2 EINECS: 212-864-8 UN: 3265 ADR: 8,II |