4-Octylbenzoic acid ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A10777.03
A10777.06
A10777.03EA
59.3
EUR
A10777.03
A10777.06
4-Octylbenzoic acid ≥99%
4-Octylbenzoic acid
Purity: 99%
CAS No.: 3575-31-3
Molecular Formula: C15H22O2
Formula Weight: 234.34
Melting Point: 97-98°
Beilstein Reference: 1954607
Formule:
C₁₅H₂₂O₂ Kookpunt: 362 °C (1013 hPa) Smeltpunt: 97…98 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00042649 CAS nummer: 3575-31-3 EINECS: 222-692-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...