α,α,α-Trifluoro-3-tolualdehyde 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23
188440050 188440250
ACRO188440050EA 59.4 EUR
ACRO188440050 ACRO188440250
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde
Formule: C₈H₅F₃O
MW: 174.12 g/mol
Kookpunt: 83…86 °C (30 mmHg)
Dichtheid: 1.3 g/cm³
Vlampunt: 68 °C
MDL Nummer: MFCD00003373
CAS nummer: 454-89-7
EINECS: 207-228-1
Order Now
Specification Test Results
Appearance Clear colorless to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4639 to 1.4659 (20°C, 589 nm)
GC ≥96.0 %