(4-Fluorophenyl)acetonitrile 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile
146720250 146721000
ACRO146720250EA 44.5 EUR
ACRO146720250 ACRO146721000
(4-Fluorophenyl)acetonitrile 99%
(4-Fluorophenyl)acetonitrile
Formule: C₈H₆FN
MW: 135.14 g/mol
Kookpunt: 119…120 °C (18 mmHg)
Smeltpunt: 86
Dichtheid: 1.126 g/cm³
Vlampunt: 108 °C
MDL Nummer: MFCD00001917
CAS nummer: 459-22-3
EINECS: 207-286-8
UN: 3276
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5020 (20°C, 589 nm)
GC ≥98.5 %