2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Fluoro-5-(trifluoromethyl)phenylisocyanate
B25630.03
B25630.06
B25630.03EA
69.2
EUR
B25630.03
B25630.06
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate ≥97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Purity: 97%
CAS No.: 69922-27-6
Molecular Formula: C8H3F4NO
Formula Weight: 205.11
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.455
Boiling Point: 53°/2.5mm
UN No.: 2206
MDL No.: MFCD00673069
Formule:
C₈H₃F₄NO Kookpunt: 53 °C (2,5 mmHg) Dichtheid: 1,418 Vlampunt: 70 °C (158 °F) |
MDL Nummer:
MFCD00673069 CAS nummer: 69922-27-6 UN: 2206 ADR: 6.1,III |