2-(Trifluoromethyl)phenyl isothiocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
α,α,α-Trifluoro-o-tolyl isothiocyanate
L10586.03
L10586.06
L10586.03EA
28.4
EUR
L10586.03
L10586.06
2-(Trifluoromethyl)phenyl isothiocyanate ≥98%
2-(Trifluoromethyl)phenyl isothiocyanate
Purity: 98%
CAS No.: 1743-86-8
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point: 105°(221°F)
Refractive Index: 1.5660
Odour: Pungent
Boiling Point: 216°
Beilstein Reference: 2693552
UN No.: 2922
MDL No.: MFCD00039644
Formule:
C₈H₄F₃NS Kookpunt: 216 °C Dichtheid: 1,346 Vlampunt: 105 °C (221 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00039644 CAS nummer: 1743-86-8 ADR: 8,III |