3-(Trifluoromethyl)phenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: α,α,α-Trifluoro-m-tolyl isothiocyanate

SB01602.1 SB01602.10 SB01602.50
MAYBSB01602.1EA 0 EUR
MAYBSB01602.1 MAYBSB01602.10 MAYBSB01602.50
3-(Trifluoromethyl)phenyl isothiocyanate
3-(Trifluoromethyl)phenyl isothiocyanate
Formule: C₈H₄F₃NS
MW: 203.19 g/mol
Kookpunt: 206…208 °C (1013 hPa)
Dichtheid: 1.33 g/cm³ (20 °C)
MDL Nummer: MFCD00004806
CAS nummer: 1840-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR