α,α,α-Trifluoro-m-toluidine 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
103190050 103191000 103195000
ACRO103190050EA 22.9 EUR
ACRO103190050 ACRO103191000 ACRO103195000
α,α,α-Trifluoro-m-toluidine 98%
α,α,α-Trifluoro-m-toluidine
Formule: C₇H₆F₃N
Kookpunt: 187…193 °C (760 mmHg)
Smeltpunt: 5…6 °C
Dichtheid: 1,303 g/cm³
Vlampunt: 85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007797
CAS nummer: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
GC ≥97.5 %