3-Fluoro-p-toluidine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01241.1
SB01241.10
SB01241.50
MAYBSB01241.1EA
0
EUR
MAYBSB01241.1
MAYBSB01241.10
MAYBSB01241.50
3-Fluoro-p-toluidine
3-Fluoro-p-toluidine
Formule:
C₇H₈FN Kookpunt: 97 °C (15 mmHg) Smeltpunt: 37 °C Dichtheid: 1,09 g/cm³ (20 °C) |
MDL Nummer:
MFCD00007762 CAS nummer: 452-77-7 EINECS: 207-212-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...