α,α,α-Trifluoro-p-toluidine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A
SB00719DA
SB00719EA
SB00719.50
MAYBSB00719DAEA
0
EUR
MAYBSB00719DA
MAYBSB00719EA
MAYBSB00719.50
α,α,α-Trifluoro-p-toluidine
α,α,α-Trifluoro-p-toluidine
Formule:
C₇H₆F₃N Kookpunt: 190 °C (1013 hPa) Smeltpunt: 3…8 °C Dichtheid: 1,3 g/cm³ (20 °C) Vlampunt: 86 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00064396 CAS nummer: 455-14-1 EINECS: 207-236-5 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...