2-Nitro-4-(trifluoromethyl)aniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB07079.1 BTB07079.10 BTB07079.50
MAYBBTB07079.1EA 0 EUR
MAYBBTB07079.1 MAYBBTB07079.10 MAYBBTB07079.50
2-Nitro-4-(trifluoromethyl)aniline
2-Nitro-4-(trifluoromethyl)aniline
Formule: C₇H₅F₃N₂O₂
Smeltpunt: 107 °C
MDL Nummer: MFCD00007155
CAS nummer: 400-98-6
EINECS: 206-926-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR