2,3,4,5,6-Pentafluorobenzyl alcohol
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
AC18836.1
AC18836EA
AC18836EE
MAYBAC18836.1EA
0
EUR
MAYBAC18836.1
MAYBAC18836EA
MAYBAC18836EE
2,3,4,5,6-Pentafluorobenzyl alcohol
2,3,4,5,6-Pentafluorobenzyl alcohol
Formule:
C₇H₃F₅O Kookpunt: 181 °C (1013 hPa) Smeltpunt: 30…32 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004602 CAS nummer: 440-60-8 EINECS: 207-126-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...