3,4-Difluorobenzyl alcohol
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
SB01790.1
SB01790.10
MAYBSB01790.1EA
0
EUR
MAYBSB01790.1
MAYBSB01790.10
3,4-Difluorobenzyl alcohol
3,4-Difluorobenzyl alcohol
Formule:
C₇H₆F₂O Kookpunt: 197 °C (1013 hPa) Smeltpunt: 9 °C Dichtheid: 1,26 g/cm³ (20 °C) |
MDL Nummer:
MFCD00010628 CAS nummer: 85118-05-4 EINECS: 285-657-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...