N,N-Dimethyl-m-toluïdine ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A13326.06
A13326.14
A13326.22
A13326.06EA
23.9
EUR
A13326.06
A13326.14
A13326.22
N,N-Dimethyl-m-toluïdine ≥98%
N,N-Dimethyl-m-toluïdine
Purity: 98%
CAS No.: 121-72-2
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 85°(185°F)
Refractive Index: 1.5500
Melting Point: -15°
Boiling Point: 212-215°
Beilstein Reference: 1422766
UN No.: 1708
MDL No.: MFCD00008305
Formule:
C₉H₁₃N Kookpunt: 212…215 °C Smeltpunt: –15 °C Dichtheid: 0,932 Vlampunt: 85 °C (185 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008305 CAS nummer: 121-72-2 EINECS: 204-495-6 UN: 1708 ADR: 6.1,II |