Benzyl isothiocyanate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Benzylisothiocyanate
402130250 402130050 402131000
ACRO402130250EA 90.5 EUR
ACRO402130250 ACRO402130050 ACRO402131000
Benzyl isothiocyanate 98%
Benzyl isothiocyanate
Formule: C₈H₇NS
Kookpunt: 243 °C (1013 hPa)
Smeltpunt: 24 °C
Dichtheid: 1,12 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004819
CAS nummer: 622-78-6
EINECS: 210-753-9
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6010 to 1.6040 (20°C, 589 nm)
GC ≥97.5 %