2-Isopropylphenyl isothiocyanate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01626DA SB01626EA SB01626.50
MAYBSB01626DAEA 0 EUR
MAYBSB01626DA MAYBSB01626EA MAYBSB01626.50
2-Isopropylphenyl isothiocyanate
2-Isopropylphenyl isothiocyanate
Formule: C₁₀H₁₁NS
Kookpunt: 82 °C (1013 hPa)
Dichtheid: 1,05 g/cm³ (20 °C)
MDL Nummer: MFCD00022053
CAS nummer: 36176-31-5