p-Tolyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Methylphenyl isothiocyanate

TL00540.1 TL00540.50
MAYBTL00540.1EA 0 EUR
MAYBTL00540.1 MAYBTL00540.50
p-Tolyl isothiocyanate
p-Tolyl isothiocyanate
Formule: C₈H₇NS
Kookpunt: 237 °C (1013 hPa)
Smeltpunt: 27…30 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004813
CAS nummer: 622-59-3
EINECS: 210-745-5
UN: 2923
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR