Phenethylamine 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: PEA, β-Phenylethylamine, 2-Phenylethylamine

A13402.AD A13402.AK A13402.AU
A13402.ADEA 25.2 EUR
A13402.AD A13402.AK A13402.AU
Phenethylamine 99%
Phenethylamine

Purity: 99% CAS No.: 64-04-0 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 81°(178°F) Refractive Index: 1.5335 Odour: Amine-like pH: 11.5 Melting Point: -60° Boiling Point: 200-202° Beilstein Reference: 507488 Merck Reference: 14,7225 UN No.: 2922 MDL No.: MFCD00008184

Formule: C₆H₅CH₂CH₂NH₂
Kookpunt: 200…202 °C
Smeltpunt: –60 °C
Dichtheid: 0,964
Vlampunt: 81 °C (178 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008184
CAS nummer: 64-04-0
EINECS: 200-574-4
UN: 2922
ADR: 8,II
Merck Index: 14,7225

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR