m-Toluidine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline
M-Clarity™ quality level = MQ200
8.08314.0100 8.08314.1000
8.08314.0100EA 35 EUR
8.08314.0100 8.08314.1000
m-Toluidine for synthesis, Sigma-Aldrich®
m-Toluidine
Formule: C₇H₉N
Kookpunt: 203 °C (1013 hPa)
Smeltpunt: –30 °C
Dichtheid: 0,98444 g/cm³ (20 °C)
Vlampunt: 90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007808
CAS nummer: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1,II
Merck Index: 13,09614
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -30 °C
Molar Mass 107.15 g/mol
Boiling Point 204 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 90 °C
Refractive Index 1.5669 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 500 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials