3-Nitro-o-toluidine 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3-Nitro-o-methylaniline, 3-Nitro-2-methylaniline, 2-Methyl-3-nitroaniline, 2-Amino-6-nitrotoluene
152020250 152021000
ACRO152020250EA 46.9 EUR
ACRO152020250 ACRO152021000
3-Nitro-o-toluidine 97%
3-Nitro-o-toluidine
Formule: C₇H₈N₂O₂
MW: 152.15 g/mol
Kookpunt: 305 °C (760 mmHg)
Smeltpunt: 87…91 °C
Dichtheid: 0.33
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007731
CAS nummer: 603-83-8
EINECS: 210-059-6
UN: 2660
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 91 °C
HPLC ≥96.0 %