3-Nitro-p-toluidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3-Nitro-4-methylaniline, 4-Methyl-3-nitroaniline, 4-Amino-2-nitrotoluene

SB01481.1 SB01481.50
MAYBSB01481.1EA 0 EUR
MAYBSB01481.1 MAYBSB01481.50
3-Nitro-p-toluidine
3-Nitro-p-toluidine
Formule: C₇H₈N₂O₂
MW: 152.15 g/mol
Kookpunt: 306
Smeltpunt: 77…78 °C
Dichtheid: 1.312 g/cm³ (20 °C)
MDL Nummer: MFCD00007910
CAS nummer: 119-32-4
EINECS: 204-314-0
UN: 2660
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR