3-Nitrobenzyl alcohol
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(3-Nitrophenyl)methanol
SB01164.1
SB01164.10
SB01164.50
MAYBSB01164.1EA
0
EUR
MAYBSB01164.1
MAYBSB01164.10
MAYBSB01164.50
3-Nitrobenzyl alcohol
3-Nitrobenzyl alcohol
Formule:
C₇H₇NO₃ Kookpunt: 317 °C (1013 hPa) Smeltpunt: 26…27 °C Dichtheid: 1,3 g/cm³ (20 °C) |
MDL Nummer:
MFCD00007273 CAS nummer: 619-25-0 EINECS: 210-588-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...