Benzylalcohol 99%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Benzenemethanol, Phenylmethanol, Phenylcarbinol
148390010 148390025 148390250
ACRO148390010EA 65.2 EUR
ACRO148390010 ACRO148390025 ACRO148390250
Benzylalcohol 99%, pure
Benzylalcohol
Formule: C₆H₅CH₂OH
MW: 108.14 g/mol
Kookpunt: 205 °C (760 mmHg)
Smeltpunt: –15 °C
Dichtheid: 1.044 g/cm³
Vlampunt: 96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004599
CAS nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5380 to 1.5410 (20°C, 589 nm)
GC ≥98.5 %