Benzylalcohol

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Benzenemethanol, Phenylmethanol, Phenylcarbinol

0336.1 0336.2 0336.3
ROTH0336.1EA 0 EUR
ROTH0336.1 ROTH0336.2 ROTH0336.3
Benzylalcohol
Benzylalcohol
Formule: C₆H₅CH₂OH
MW: 108.14 g/mol
Kookpunt: 205
Smeltpunt: –15.3 °C
Dichtheid: 1.05 g/cm³ (20 °C)
Vlampunt: 101
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004599
CAS nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR