4-Bromobenzophenone 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
106720250 106721000
ACRO106720250EA 34.9 EUR
ACRO106720250 ACRO106721000
4-Bromobenzophenone 97%
4-Bromobenzophenone
Formule: C₁₃H₉BrO
Kookpunt: 350 °C (1013 hPa)
Smeltpunt: 78…82 °C
MDL Nummer: MFCD00000103
CAS nummer: 90-90-4
EINECS: 202-024-9
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥98.5 %
Color scale ≤50 APHA