Dibenzylamine ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D0152-25G D0152-500G
TCIAD0152-25GEA 17.6 EUR
TCIAD0152-25G TCIAD0152-500G
Dibenzylamine ≥97.0% (by GC)
Dibenzylamine
Formule: C₆H₅CH₂NHCH₂C₆H₅
Kookpunt: 270°C
Smeltpunt: –26°C
Dichtheid: 1,03 g/cm³ (20 °C)
Vlampunt: 150°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004770
CAS nummer: 103-49-1
EINECS: 203-117-7
ADR: 6.1,III
Merck Index: 14,03011
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45904366b8ca909d91a366def76ade7c88380370&vid=45904366b8ca909d91a366def76ade7c88380370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR