Diphenylmethane ≥99%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 99+%
CAS No.: 101-81-5
Molecular Formula: (C6H5)2CH2
Formula Weight: 168.24
Flash Point: 130°(266°F)
Refractive Index: 1.5770
Melting Point: 22-26°
Boiling Point: 262-264°
Beilstein Reference: 1904982
Merck Reference: 14,3328
UN No.: 3077
MDL No.: MFCD00004781
Formule:
C₁₃H₁₂ Kookpunt: 262…264 °C Smeltpunt: 22…26 °C Dichtheid: 1,010 Vlampunt: 130 °C (266 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004781 CAS nummer: 101-81-5 EINECS: 202-978-6 UN: 3077 ADR: 9,III Merck Index: 14,3328 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...