4-Pentylbiphenyl
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB10236.1
BTB10236.10
BTB10236.50
MAYBBTB10236.1EA
0
EUR
MAYBBTB10236.1
MAYBBTB10236.10
MAYBBTB10236.50
4-Pentylbiphenyl
4-Pentylbiphenyl
Formule:
C₁₇H₂₀ Kookpunt: 106…109 °C (0,1 mmHg) Dichtheid: ∼0,94 g/cm³ (20 °C) Vlampunt: >110 °C |
MDL Nummer:
MFCD00009503 CAS nummer: 7116-96-3 EINECS: 230-421-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...