3,4-Dimethylbenzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00319.1 TL00319.10 TL00319.50
MAYBTL00319.1EA 0 EUR
MAYBTL00319.1 MAYBTL00319.10 MAYBTL00319.50
3,4-Dimethylbenzamide
3,4-Dimethylbenzamide
Formule: C₉H₁₁NO
MW: 149.19 g/mol
MDL Nummer: MFCD00052960
CAS nummer: 5580-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR